Learn More
Gilson™ SPE Manifold
SPE Manifold Kit
Brand:  Gilson™ 37417011
Additional Details : Weight : 0.45000kg
Product Code. 16686165
  • € 2157.10 / Each
Estimated Shipment: 13-06-2024
to see stock.
Add to basket
Description
Description
  • Cartridge ejection plate (quantity 1)
  • Cartridge rack, SPE, 24-positions, 1 mL (quantity 1)
  • Gasket with adhesive-backed seal, 24-positions (quantity 2)
  • Manifold, SPE, 24-positions (quantity 1)
  • O-ring manifold seal (quantity 1)
  • Tube rack, SPE collection, 1–3 mL, 12 x 75 mm (quantity 1)
Specifications
Specifications
24 Position Kit
1 mL
SPE Manifold