Learn More
Spebrutinib, MedChemExpress
Description
Spebrutinib (AVL-292
Specifications
Specifications
| Chemical Name or Material | Spebrutinib |
| CAS | 1202757-89-8 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H22FN5O3 |
| Quantity | 100 mg |
| Synonym | AVL-292 CC-292 |
| Solubility Information | DMSO : ≥ 45 mg/mL (106.27 mM) |
| SMILES | COCCOC1=CC=C(NC2=NC=C(F)C(NC3=CC=CC(NC(C=C)=O)=C3)=N2)C=C1 |
| Molecular Weight (g/mol) | 423.44 |
| Show More |
Research purposes only