missing translation for 'onlineSavingsMsg'
Learn More

Spebrutinib, MedChemExpress

Product Code. 16459927
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16459927 5 mg 5mg
16477416 100 mg 100mg
16467448 10 mg 10mg
16413477 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16459927 Supplier Medchem Express Supplier No. HY180125mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Spebrutinib (AVL-292; CC-292) is a covalent, orally active, and highly selective with an IC50 of 0.5 nM.

Spebrutinib (AVL-292

Specifications

Chemical Name or Material Spebrutinib
CAS 1202757-89-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H22FN5O3
Quantity 5 mg
Synonym AVL-292 CC-292
Solubility Information DMSO : ≥ 45 mg/mL (106.27 mM)
SMILES COCCOC1=CC=C(NC2=NC=C(F)C(NC3=CC=CC(NC(C=C)=O)=C3)=N2)C=C1
Molecular Weight (g/mol) 423.44
Formula Weight 423.44
Percent Purity 95.15%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.