missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPECC1L Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94163-0.1ml
This item is not returnable.
View return policy
Description
SPECC1L Polyclonal antibody specifically detects SPECC1L in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SPECC1L | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| Cytokinesis And Spindle Organization A, Cytospin A, CYTSA, GBBB2, KIAA0376, OBLFC1, Renal Carcinoma Antigen NY-REN-22, SPECC1-Like, SPECC1-Like Protein, Sperm Antigen With Calponin Homology And Coiled-Coil Domains 1-Like | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 200-500 of human SPECC1L (NP_056145.4). LHLRNELRDMRAQLGINEDHSEGDEKSEKETIMAHQPTDVESTLLQLQEQNTAIREELNQLKNENRMLKDRLNALGFSLEQRLDNSEKLFGYQSLSPEITPGNQSDGGGTLTSSVEGSAPGSVEDLLSQDENTLMDHQHSNSMDNLDSECSEVYQPLTSSDDALDAPSSSESEGIPSIERSRKGSSGNASEVSVACLTERIHQMEENQHSTSEELQATLQELADLQQITQELNSENERLGEEKVILMESLCQQSDKLEHFSRQIEYFRSLLDEHHISYVIDEDVKSGRYMELEQRYMDLAE | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Cell Cycle and Replication, Immunology | |
| 23384 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction