missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrin alpha 1 Antibody (SR1447), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22179-100ul
This item is not returnable.
View return policy
Description
Spectrin alpha 1 Monoclonal antibody specifically detects Spectrin alpha 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
Specifications
| Spectrin alpha 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| SR1447 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| alpha-I spectrin, EL2, erythrocyte, HS3, spectrin, alpha, erythrocytic 1 (elliptocytosis 2) | |
| A synthesized peptide derived from human Spectrin alpha 1 (Uniprot #: P02549) | |
| 100 μg | |
| Cell Biology, Cellular Markers | |
| 6708 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction