missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrin Alpha 1 (Erythrocyte Marker) Mouse, Clone: SPTA1/1810, enQuireBio™
Mouse Monoclonal Antibody
€ 199.00 - € 510.00
Specifications
Antigen | Spectrin Alpha 1 (Erythrocyte Marker) |
---|---|
Clone | SPTA1/1810 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15940908
|
EnquireBio
6708-MSM1-P1 |
100 μg |
€ 510.00
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15930908
|
EnquireBio
6708-MSM1-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Spectrin Alpha 1 (Erythrocyte Marker) | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant human SPTA fragment (apprx. aa356-475) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
240kDa |
SPTA1/1810 | |
Immunohistochemistry, ELISA | |
Unconjugated | |
RUO | |
6708 | |
IgG1 κ | |
Protein A/G purified from Bioreactor concentrate. | |
Cytoplasmic and Cell Surface | |
Jurkat cells. Placenta or Tonsil. |