missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrin beta 1 Antibody (RG/26), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08440AF647
This item is not returnable.
View return policy
Description
Spectrin beta 1 Monoclonal antibody specifically detects Spectrin beta 1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Spectrin beta 1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| beta-I spectrin, EL3, erythrocyte, HS2, HSPTB1, membrane cytoskeletal protein, spectrin, beta, erythrocytic, SPTB1 | |
| Red cell membranes | |
| 100 μg | |
| Cell Biology, Cellular Markers | |
| 6710 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| RG/26 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction