missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrin beta 3 Antibody (SPTBN2/1582), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54488MFV500
This item is not returnable.
View return policy
Description
Spectrin beta 3 Monoclonal antibody specifically detects Spectrin beta 3 in Human samples. It is validated for Flow Cytometry
Specifications
| Spectrin beta 3 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| SPTBN2/1582 | |
| Flow Cytometry | |
| Beta-III spectrin, glutamate transporter EAAT4-associated protein 41, GTRAP41, KIAA0302, SCA5, spectrin beta chain, brain 2, spectrin, beta, non-erythrocytic 2, Spectrin, non-erythroid beta chain 2, spinocerebellar ataxia 5 | |
| Recombinant fragment (around aa356-475) of human Spectrin beta 3 protein (exact sequence is proprietary) (Uniprot: O15020) | |
| 0.1 mL | |
| Neuroscience | |
| 6712 | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction