Learn More
Invitrogen™ Spectrin beta-3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560406
Description
Immunogen sequence: RLWRFLWEVG EAEAWVREQQ HLLASADTGR DLTGALRLLN KHTALRGEMS GRLGPLKLTL EQGQQLVAEG HPGASQASA Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
Spectrins are principle components of a cell's membrane-cytoskeleton and are composed of two alpha and two beta spectrin subunits. The protein encoded by this gene (SPTBN2), is called spectrin beta non-erythrocytic 2 or beta-III spectrin. It is related to, but distinct from, the beta-II spectrin gene which is also known as spectrin beta non-erythrocytic 1 (SPTBN1). SPTBN2 regulates the glutamate signaling pathway by stabilizing the glutamate transporter EAAT4 at the surface of the plasma membrane. Mutations in this gene cause a form of spinocerebellar ataxia, SCA5, that is characterized by neurodegeneration, progressive locomotor incoordination, dysarthria, and uncoordinated eye movements.
Specifications
| Spectrin beta-3 | |
| Polyclonal | |
| Unconjugated | |
| Sptbn2 | |
| beta SpIII sigma 1; beta-III spectrin; beta-spectrin 3; glutamate transporter EAAT4-associated protein 41; GTRAP41; KIAA0302; mKIAA0302; SCA5; SCAR14; Spectrin Beta; spectrin beta 3; spectrin beta chain, brain 2; spectrin beta chain, non-erythrocytic 2; spectrin beta III sigma 2; spectrin beta, non-erythrocytic 2; spectrin, beta, non-erythrocytic 2; spectrin, non-erythroid beta chain 2; spectrin-like protein GTRAP41; spinocerebellar ataxia 5 protein; Spnb3; SPNB-3; SPTBN2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6712 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15020 | |
| Sptbn2 | |
| Recombinant protein corresponding to Human Spectrin beta-3. Recombinant protein control fragment (Product #RP-103863). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.