missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrometric Oil Reference Standard D19-5: Ag, Al, B, Ba, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, Pb, Si, Sn, Ti, V, Zn @ 5 μg/g in Aviation Reference Oil, VHG™
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Spectrometric Oil Reference Standard D19-5: Ag, Al, B, Ba, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, Pb, Si, Sn, Ti, V, Zn @ 5 μg/g in Aviation Reference Oil
Specifications
Specifications
| Chemical Name or Material | Aluminium/Barium/Boron/Cadmium/Chromium/Copper/Iron/Lead/Magnesium/Manganese/Molybdenum/Nickel/Silicon/Silver/Sodium/Tin/Titanium/Vanadium/Zinc |
| CAS | 7429-90-5, 7440-39-3, 7440-42-8, 7440-43-9, 7440-47-3, 7440-50-8 |
| Methods | RDE |
| Analyte or Component Names | Aluminium/Barium/Boron/Cadmium/Chromium/Copper/Iron/Lead/Magnesium/Manganese/Molybdenum/Nickel/Silicon/Silver/Sodium/Tin/Titanium/Vanadium/Zinc |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Aviation Oil |
| Quantity | 100 g |
| Concentration or Composition (by Analyte or Components) | Aluminium 5 μg/g/Barium 5 μg/g/Boron 5 μg/g/Cadmium 5 μg/g/Chromium 5 μg/g/Copper 5 μg/g/Iron 5 μg/g/Lead 5 μg/g/Magnesium 5 μg/g/Manganese 5 μg/g/Molybdenum 5 μg/g/Nickel 5 μg/g/Silicon 5 μg/g/Silver 5 μg/g/Sodium 5 μg/g/Tin 5 μg/g/Titanium 5 μg/g/Vanadium 5 μg/g/Zinc 5 μg/g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction