Learn More
Spectrum™ Trial Size Kits for Biotech-Grade RC Dialysis Membrane Tubing
0.5 meter (1.6 ft) dialysis tubing length ideal for small applications or membrane evaluation
Brand: Spectrum™ 133 270T
Additional Details : Weight : 0.05000kg
Includes: 0.5m Biotech RC Tubing with glycerine, 2 Universal Tubing Closures, 5 opening picks
Description
Ultrapure Biotech RC is a premium dialysis membrane that provides superior molecular selectivity than Standard RC and superior tolerance than Biotech CE
- Properties: Hydrophilic, symmetric porosity
- 3 MWCOs: 3.5-5 kD, 8-10 kD and 20 kD
- Ultrapure, contains no heavy metals or sulfides
Specifications
Dialysis Trial Kit | |
10 mm | |
0.5m Biotech RC tubing with glycerine, 2 universal tubing closures, 5 opening picks | |
Ideal for membrane and application evaluation for more critical separations |
8 to 10 kDa | |
0.39 in. | |
Each |