missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Speedy/Ringo Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93759-0.02ml
This item is not returnable.
View return policy
Description
Speedy/Ringo Polyclonal antibody specifically detects Speedy/Ringo in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Speedy/Ringo | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| hSpy/Ringo A, MGC110856, Rapid inducer of G2/M progression in oocytes A, RINGO A, Ringo3, SPDY1, speedy homolog 1 (Drosophila), speedy homolog A (Xenopus laevis), speedy protein A, speedy-1, Spy1, SPY1MGC57218 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-313 of human SPDYA (NP_877433.2). MRHNQMCCETPPTVTVYVKSGSNRSHQPKKPITLKRPICKDNWQAFEKNTHNNNKSKRPKGPCLVIQRQDMTAFFKLFDDDLIQDFLWMDCCCKIADKYLLAMTFVYFKRAKFTISEHTRINFFIALYLANTVEEDEEETKYEIFPWALGKNWRKLFPNFLKLRDQLWDRIDYRAIVSRRCCEEVMAIAPTHYIWQRERSVHHSGAVRNYNRDEVQLPRGPSATPVDCSLCGKKRRYVRLGLSSSSSLSSHTAGVTEKHSQDSYNSLSMDIIGDPSQAYTGSEVVNDHQSNKGKKTNFLKKDKSMEWFTGSEE | |
| 0.02 mL | |
| Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair | |
| 245711 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction