missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPEF2 Polyclonal Antibody

SPEF2 Polyclonal Antibody
Brand: Thermo Scientific PA559141
This item is not returnable.
View return policy
Description
SPEF2 Polyclonal Antibody for IHC
Specifications
| SPEF2 | |
| Polyclonal | |
| Unconjugated | |
| SPEF2 | |
| C230086A09Rik; cancer/testis antigen 122; CT122; KIAA1770; Kpl2; Protein KPL2; SPEF2; sperm flagellar 2; sperm flagellar protein 2; testis tissue sperm-binding protein Li 47a | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 79925 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9C093 | |
| SPEF2 | |
| Recombinant protein corresponding to Human SPEF2. Recombinant protein control fragment (Product #RP-102886). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction