missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SPEF2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15796721 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15796721 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15796721 Supplier Invitrogen™ Supplier No. PA558854

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VPQPPKPGSE EWVYVNEPVP EEMPLFLVPY WELIENSYIN TIKTVLRHLR EDQHTVLAYL YEIRTSFQEF LKRPDHKQDF VAQWQ Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 78%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen SPEF2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPEF2
Gene Accession No. Q9C093
Gene Alias C230086A09Rik; cancer/testis antigen 122; CT122; KIAA1770; Kpl2; Protein KPL2; SPEF2; sperm flagellar 2; sperm flagellar protein 2; testis tissue sperm-binding protein Li 47a
Gene Symbols SPEF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPEF2. Recombinant protein control fragment (Product #RP-96899).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79925
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.