missing translation for 'onlineSavingsMsg'
Learn More

SPEN Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB100-58799JF646

Product Code. 18742230

  • ONLINE EXCLUSIVE PRICE
    € 681.00
    € 643.65 / 0.10mL
    Save € 37.35
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SPEN Polyclonal antibody specifically detects SPEN in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SPEN
Polyclonal
Western Blot
HIAA0929, KIAA0929nuclear receptor transcription cofactor, MINTMsx2 interacting nuclear target (MINT) homolog, RBM15C, SHARPmsx2-interacting protein, SMART/HDAC1 associated repressor protein, SMART/HDAC1-associated repressor protein, SPEN homolog, spen homolog, transcriptional regulator (Drosophila)
The immunogen recognized by this antibody maps to a region between residue 3375 and 3425 of human SMART/HDAC1 associated repressor protein (spen homolog, transcriptional regulator) using the numbering given in entry NP_055816.2 (GeneID 23013).
0.1 mL
Primary
Human, Mouse
Purified
Western Blot
Janelia Fluor 646
50mM Sodium Borate
Rabbit
Immunogen affinity purified
RUO
23013
Store at 4C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.