Learn More
SPEN Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-58799PCP
Product Code. 18507371
  • € 661.00 / 0.10mL
Description
SPEN Polyclonal antibody specifically detects SPEN in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation, Proximity Ligation Assay, ChIP assay (ChIP)