missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spermatogensis associated protein 1 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41142G
This item is not returnable.
View return policy
Description
Spermatogensis associated protein 1 Polyclonal antibody specifically detects Spermatogensis associated protein 1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Spermatogensis associated protein 1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| SP-2, SPAP1, SPATA1 spermatogenesis associated 1 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human SPATA1. The immunogen is located within amino acids 300 - 350 of SPATA1. Amino Acid Squence: AYNGWKKKYLETKKVTAS | |
| 0.1 mL | |
| Signal Transduction | |
| 100505741 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction