Learn More
Spermine synthase Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NB100-75148-0.025mg
This item is not returnable.
View return policy
Description
Spermine synthase Polyclonal specifically detects Spermine synthase in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Spermine synthase | |
| Polyclonal | |
| Western Blot 1:100-1:2000 | |
| P52788 | |
| SMS | |
| The immunogen for this antibody was a sequence from 206a.a. to 335a.a. from Spermine synthase(a 368a.a. long protein) | |
| 0.025 mg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6611 | |
| Human |
| Western Blot | |
| Unconjugated | |
| PBS with No Preservative | |
| EC 2.5.1.22, MRSRSpS, Snyder-Robinson X-linked mental retardation syndrome, Spermidine aminopropyltransferase, spermine synthase, SPMSYSRS | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Spermine synthase | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |