Learn More
Spermine synthase Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand:  Novus Biologicals NB100-75148-0.025mg
 View more versions of this product
Product Code. 18503604
  • € 370.00 / 25µg
Description
Spermine synthase Polyclonal specifically detects Spermine synthase in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Spermine synthase
Polyclonal
Western Blot 1:100-1:2000
P52788
SMS
The immunogen for this antibody was a sequence from 206a.a. to 335a.a. from Spermine synthase(a 368a.a. long protein)
0.025 mg
Core ESC Like Genes, Stem Cell Markers
6611
Human
Western Blot
Unconjugated
PBS with No Preservative
EC 2.5.1.22, MRSRSpS, Snyder-Robinson X-linked mental retardation syndrome, Spermidine aminopropyltransferase, spermine synthase, SPMSYSRS
Chicken
Affinity Purified
RUO
Primary
Spermine synthase
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.