Learn More
SPEX CertiPrep™ Method AOAC 2007.01 SPEXQuE™ QuEChERS Kit
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
15 mL
Unit Size:
Pack of 50
Description
Meeting all your QuEChERS needs with general purpose kits suitable for removal of polar organic acids, some low to moderate levels of sugars and lipids.
- US (AOAC Method 2007.01)
Specifications
Specifications
| Packaging | 50 pk. |
| Methods | AOAC 2007.01 |
| Analyte or Component Names | 400mg PSA, 1200mg MgSO4 |
| Technique | Pesticide Extraction |
| Type | PSA Dispersive SPE Clean-up Tube |
| Element | PSA, MgSO4 |
| Quantity | 15 mL |