Learn More
SPEX CertiPrep™ Method AOAC 2007.01 SPEXQuE™ QuEChERS Kit
Sensible Solutions for Pesticide Extraction
Brand:  SPEX Certiprep AOAC-PSA-15ML
Additional Details : Weight : 0.09000kg
 View more versions of this product
Product Code. 15255866
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Meeting all your QuEChERS needs with general purpose kits suitable for removal of polar organic acids, some low to moderate levels of sugars and lipids.
  • US (AOAC Method 2007.01)
Specifications
Specifications
50 pk.
PSA Dispersive SPE Clean-up Tube
400mg PSA, 1200mg MgSO4
15 mL
AOAC 2007.01
Pesticide Extraction
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers