Learn More
Cole-Parmer™ Replacement Impactor for Large Grinding Vial Set (61043-09), Stainless Steel
Product Code. 12534196 Shop All SPEX Certiprep Products
Click to view available options
Packaging Quantity:
1 Ea.
Unit Size:
Each
Product Code. 12534196
Brand: Cole-Parmer™ 6801P
This item requires a price update prior to being ordered.
Request a Price for this item here.
Replacement impactor for 6801 Large Grinding Vial Set.
  • Made of 440C stainless steel.
  • Withstands liquid nitrogen temperatures.
  • Autoclavable and reusable.
Specifications
Product Type Impactor
Autoclavable Autoclavable
Disposable Reusable
For Use With (Equipment) 6801 Large Grinding Vial Set (61043-09) of SpexSamplePrep Large-Capacity Freezer/Mill
Material Stainless Steel (440 C)
Packaging Quantity 1 Ea.