Learn More
Cole-Parmer™ Replacement Impactor for Large Grinding Vial Set (61043-09), Stainless Steel
Shop All SPEX Certiprep Products
Click to view available options
Packaging Quantity:
1 Ea.
Unit Size:
Each
Description
- Made of 440C stainless steel.
- Withstands liquid nitrogen temperatures.
- Autoclavable and reusable.
Specifications
Specifications
| Product Type | Impactor |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| For Use With (Equipment) | 6801 Large Grinding Vial Set (61043-09) of SpexSamplePrep Large-Capacity Freezer/Mill |
| Material | Stainless Steel (440 C) |
| Packaging Quantity | 1 Ea. |