Learn More
Cole-Parmer™ XRF Prep-Aid™ Grinding Aids and Sample Binders
Product Code. 15380552 Shop All SPEX Certiprep Products
Change view
Click to view available options
Type:
Paraffin Binder
Quantity:
450 g
Unit Size:
450g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Quantity unitSize
15380552 Paraffin Binder 450 g 450g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15380552 Supplier Cole-Parmer™ Supplier No. 3646450
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
Prepare samples for reproducible and contamination-free results with XRF Prep-Aid™ Grinding Aids and Sample Binders.
Ideal for samples which crumble or ablate after pressing.
  • Select binders from powder, tablet, or liquid form depending on your application.
  • High-quality sample preparation is the key to achieving consistent and reliable analytical results.
  • Powders are ideal for samples that crumble or ablate after pressing.
  • They blend quickly and completely with samples to form XRF (x-ray fluorescence) sample disks.
  • Tablets are an efficient way to add binder to XRF sample disks.
  • Liquid easily mixes with the sample.
Specifications
Type Paraffin Binder
For Use With (Application) Samples Which Crumble or Ablate After Pressing
Particle Size ≤20 μm
Quantity 450 g