missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPG20 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142444
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.
SPG20 is a protein containing a MIT (Microtubule Interacting and Trafficking molecule) domain, and is implicated in regulating endosomal trafficking and mitochondria function. The protein localizes to mitochondria and partially co-localizes with microtubules. Stimulation with epidermal growth factor (EGF) results in protein translocation to the plasma membrane, and the protein functions in the degradation and intracellular trafficking of EGF receptor.
Specifications
| SPG20 | |
| Polyclonal | |
| Unconjugated | |
| SPART | |
| AI840044; C79168; KIAA0610; mKIAA0610; Spart; Spartin; Spartin-like protein; spastic paraplegia 20 (Troyer syndrome); spastic paraplegia 20 (Troyer syndrome) homolog; Spastic paraplegia 20 protein; spastic paraplegia 20, spartin (Troyer syndrome) homolog; spastic paraplegia 20, spartin (Troyer syndrome) homolog (human); SPG20; TAHCCP1; Trans-activated by hepatitis C virus core protein 1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 23111 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8N0X7 | |
| SPART | |
| Peptide with sequence C-EASGTDVKQLDQGNK. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction