missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPG7 Antibody (OTI1G4), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74323B
This item is not returnable.
View return policy
Description
SPG7 Monoclonal specifically detects SPG7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| SPG7 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| SPG7 | |
| Full length recombinant protein of human SPG7 (NP_003110) produced in E.coli. | |
| 0.1 mL | |
| 6687.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1G4 | |
| Western Blot, Immunohistochemistry | |
| CARMGC126331, cell matrix adhesion regulator, CMARMGC126332, EC 3.4.24, EC 3.4.24.-, FLJ37308, paraplegin, PGNcell adhesion regulator, spastic paraplegia 7 (pure and complicated autosomal recessive), Spastic paraplegia 7 protein, SPG5C | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction