missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPG7 Monoclonal Antibody (OTI7D10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800012S
This item is not returnable.
View return policy
Description
SPG7 Monoclonal antibody specifically detects SPG7 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| SPG7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UQ90 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6687 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI7D10 | |
| Unconjugated | |
| SPG7 | |
| AI452278; AU015315; CAR; cell adhesion regulator; cell matrix adhesion regulator; CMAR; paraplegin; paraplegin, isoform 1; PGN; spastic paraplegia 7 (pure and complicated autosomal recessive); spastic paraplegia 7 homolog (human); spastic paraplegia 7 protein; SPG5C; SPG7; SPG7 matrix AAA peptidase subunit, paraplegin; SPG7, paraplegin matrix AAA peptidase subunit | |
| Human recombinant protein fragment corresponding to amino acids 300-573 of human SPG7 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction