Learn More
SPG7 Mouse anti-Human, Clone: OTI3H10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF800010
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Putative ATP-dependent zinc metalloprotease.Specifications
| SPG7 | |
| Monoclonal | |
| Unconjugated | |
| SPG7 | |
| CAR, CMAR, PGN, SPG5C | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6687 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI3H10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9UQ90 | |
| SPG7 | |
| Human recombit protein fragment corresponding to amino acids 300-573 of human SPG7 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.