missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPG7 Recombinant Rabbit Monoclonal Antibody (HL2236)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550450
This item is not returnable.
View return policy
Description
SPG7 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a mitochondrial metalloprotease protein that is a member of the AAA family. Members of this protein family share an ATPase domain and have roles in diverse cellular processes including membrane trafficking, intracellular motility, organelle biogenesis, protein folding, and proteolysis. Mutations in this gene cause autosomal recessive spastic paraplegia 7. Two transcript variants encoding distinct isoforms have been identified.
Specifications
| SPG7 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q3ULF4, Q7TT47, Q9UQ90 | |
| SPG7 | |
| Recombinant fragmemt of human SPG7. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL2236 | |
| Unconjugated | |
| SPG7 | |
| AI452278; AU015315; CAR; cell adhesion regulator; cell matrix adhesion regulator; CMAR; paraplegin; paraplegin, isoform 1; PGN; spastic paraplegia 7 (pure and complicated autosomal recessive); spastic paraplegia 7 homolog (human); spastic paraplegia 7 protein; SPG5C; SPG7; SPG7 matrix AAA peptidase subunit, paraplegin; SPG7, paraplegin matrix AAA peptidase subunit | |
| Rabbit | |
| Protein A | |
| RUO | |
| 234847, 353231, 6687 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction