missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sphingosine Kinase 1/SPHK1 Antibody (OTI1A6), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74332DL594
This item is not returnable.
View return policy
Description
Sphingosine Kinase 1/SPHK1 Monoclonal antibody specifically detects Sphingosine Kinase 1/SPHK1 in Human, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Sphingosine Kinase 1/SPHK1 | |
| Monoclonal | |
| DyLight 594 | |
| EC 2.7.1.91, sphingosine kinase 1, SPHKSK 1, SPK, SPK 1 | |
| Full length recombinant protein of human Sphingosine Kinase 1/SPHK1 (NP_068807) produced in HEK293T cells. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 8877 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| OTI1A6 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Rat, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction