missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SPHK1 Recombinant Rabbit Monoclonal Antibody (2L6P4)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA551350

Product Code. 18066081

  • € 456.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MDPAGGPRGV LPRPCRVLVL LNPRGGKGKA LQLFRSHVQP LLAEAEISFT LMLTERRNHA RELVRSEELG RWDALVVMSG DGLMHEVVNG LMERPDWETA IQKPLCSLPA GSGNALAASL NHYAGYEQVT NEDLLTNCTL LLCRRLLSPM NLLSLHTASG.

SPHK1, along with SPHK2, catalyzes the phosphorylation of sphingosine producing the signaling molecule sphingosine-1-phosphate (S1P). S1P signals through G-protein coupled receptors and controls diverse cellular processes. Inhibitors of SPHKs are viewed as potential therapeutics for the treatment of cancer. SPHK1 catalyzes the phosphorylation of sphingosine to sphingosine 1 phosphate (S1P), an important signaling molecule with intra- and extracellular functions. Inside the cell S1P acts as a signaling molecule like other sphingolipid metabolites like ceramide and sphingosine. S1P has been implicated in regulating cell differentiation, calcium mobilization from intracellular stores, and apoptosis. The cell surface receptors for S1P are the EDG family of G protein-coupled receptors (S1P Receptors).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SPHK1
Recombinant Monoclonal
0.203 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Q9NYA1
SPHK1
Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human SPHK1 (NP_001136074.1).
100 μL
Primary
Human
Antibody
Liquid
ELISA, Western Blot
2L6P4
Unconjugated
SPHK1
1110006G24Rik; Acetyltransferase SPHK1; RP23-193A16.4; SK 1; Sk1; SKase; Sphingosine kinase 1; SPHK; SPHK1; SPK; SPK 1; Spk1
Rabbit
Affinity chromatography
RUO
8877
-20°C, Avoid Freeze/Thaw Cycles
Liquid
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.