Learn More
Invitrogen™ SPIB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121102
Description
Positive test controls include: Raji. The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: MLALEAAQLD GPHFSCLYPD GVFYDLDSCK HSSYPDSEGA PDSLWDWTVA PPVPATPYEA FDPAAAAFSH PQAAQLCYEP PTYSPAGNLE LAPSLEAPGP GLPAYPTENF ASQTLVPPAY APYPSPVLSE EEDLPLDSPA LEVSDSESDE ALVAGPEGKG SEAGTRKKLR LYQFLLGLLT RGDMRECVWW VEPGAGVFQF SSKHKELLAR RWGQQKGNRK RMTYQKLARA LRNYAKTGEI RKVKRKLTYQ FDSALLPAVR RA.
Spi-B is a transcription factor belonging to a subfamily of ETS transcription factors with a conserved ETS domain that mediates specific DNA binding. Spi-B may form homotypic interactions and is known to interact with IRF4, CREBBP, EP300, SPI1/PU.1 related, JUN and TBP. As a sequence specific transcriptional activator it binds to the PU-box, a purine-rich DNA sequence (5'-GAGGAA-3') that can act as a lymphoid-specific enhancer. Two isoforms of Spi-B are reported. Spi-B is usually expressed in plasmacytoid dendritic cells (pDCs), also known as type 2 DC precursors (pre-DC2) or natural interferon (IFN)-producing cells and promotes their development and block viral replication. Spi-B is not expressed in T-cells or granulocytes but may be present in stem cell populations of the liver and B-cells where it helps in B-cell receptor (BCR) signaling, normal B-cell development and antigenic stimulation.
Specifications
| SPIB | |
| Polyclonal | |
| Unconjugated | |
| SPIB | |
| B42b-like; Ets transcription factor Spi-B; Spi-1/PU.1 related; Spib; SPI-B; Spi-B transcription factor; Spi-B transcription factor (Spi-1/PU.1 related); Transcription factor Spi-B | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6689 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.75 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q01892 | |
| SPIB | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-262 of human SPIB (NP_003112.2). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.