missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SPIC Polyclonal Antibody
GREENER_CHOICE

Product Code. 15885953
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15885953 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15885953 Supplier Invitrogen™ Supplier No. PA567537

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SPIC Polyclonal Antibody for Western Blot, IHC (P)

Controls the development of red pulp macrophages required for red blood cells recycling and iron homeostasis. Transcription factor that binds to the PU-box, a purine-rich DNA sequence (5'-GAGGA[AT]-3') that can act as a lymphoid-specific enhancer. Regulates VCAM1 gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPIC
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Spic
Gene Accession No. Q6P3D7, Q8N5J4
Gene Alias AU019198; C76795; Prf; Pu.1-related factor; SPIC; SPI-C; Spi-C transcription factor; Spi-C transcription factor (Spi-1/PU.1 related); Transcription factor Spi-C
Gene Symbols Spic
Host Species Rabbit
Immunogen Full length Fusion protein of human SPIC.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121599, 20728
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.