missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SPIDR Polyclonal Antibody
GREENER_CHOICE

Product Code. 15753461 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15753461 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15753461 Supplier Invitrogen™ Supplier No. PA555541

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: ENSAKKKLLR GGLAERLNGL QNRERSAISL WRHQCISYQK TLSGRKSGVL TVKILELHEE CAMQVAMCEQ LLGSPATSSS QSVAPR Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.

Plays a role in DNA double-strand break (DBS) repair via homologous recombination (HR). Serves as a scaffolding protein that helps to promote the recruitment of DNA-processing enzymes like the helicase BLM and recombinase RAD51 to site of DNA damage, and hence contributes to maintain genomic integrity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPIDR
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPIDR
Gene Accession No. Q14159
Gene Alias DNA repair-scaffolding protein; k0146; KIAA0146; Scaffolding protein involved in DNA repair; SPIDR
Gene Symbols SPIDR
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPIDR. Recombinant protein control fragment (Product #RP-93870).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23514
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.