Learn More
New Pig™ Spill Kits
Click to view available options
Capacity (Metric):
7.1 L
15 L
79.49 L
Unit Size:
Each
Specifications
Specifications
| Color | White |
| Capacity (English) | 4 gal. |
| Capacity (Metric) | 15 L |
| Material | Polyethylene, Polypropylene, Magnesium Aluminosilicate |
| For Use With (Application) | Oils, Coolants, Solvents, Water--Universal |
| Includes | 3 in. dia. x 10 ft. L PIG™ Blue Absorbent Sock, 3 in. dia. x 48 in. L PIG™ Blue Absorbent Sock, (4) 15 in. W x 20 in. L PIG™ Absorbent Mat Pad, (6) 2 in. W x 6 in. L Tamperproof Seal Label, (2) 18 in. W x 30 in. H Polyethylene Disposal Bags, Bucket with Lid, 6.5 gallons, printed |
| Height (English) | 19.45 in. |
| Diameter (English) | 12.69 in. |
| Weight (English) | 11 lb. |