missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brookfield™ Spindle LV-3
Click to view available options
Viscosity Range:
200 to 400,000 cP
Unit Size:
Each
Description
- Made of 302 grade stainless steel
- For use with standard LV torque Brookfield viscometers and rheometers
Specifications
Specifications
| Viscosity Range | 200 to 400,000 cP |
| For Use With (Equipment) | Rotational Viscometer Models 98945-00/98945-05 |
| Material | Stainless Steel (302) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction