missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPINK4 Antibody (248), DyLight 405 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89953V
This item is not returnable.
View return policy
Description
SPINK4 Monoclonal antibody specifically detects SPINK4 in Human samples. It is validated for ELISA
Specifications
| SPINK4 | |
| Monoclonal | |
| DyLight 405 | |
| gastrointestinal peptide, MGC133107, serine peptidase inhibitor, Kazal type 4, serine protease inhibitor Kazal-type 4, serine protease inhibitor, Kazal type 4 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human SPINK4 (NP_055286.1; Met1-Cys86). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 248 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 27290 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction