Learn More
SPINK7 Antibody, Biotin, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-41141B
This item is not returnable.
View return policy
Description
SPINK7 Polyclonal antibody specifically detects SPINK7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SPINK7 | |
| Polyclonal | |
| PBS | |
| Chicken | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Biotin | |
| ECG2MGC133106, ECRG2, ECRG-2, esophagus cancer related gene 2, Esophagus cancer-related gene 2 protein, esophagus cancer-related gene-2, MGC133105, serine peptidase inhibitor, Kazal type 7 (putative), serine protease inhibitor Kazal-type 7 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human ECRG2. The immunogen is located within the last 50 amino acids of ECRG2. Amino Acid Squence: TESLKSNGRVQFLHD | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 84651 | |
| Store at 4°C in the dark. | |
| IgY |