missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPINT1 Monoclonal Antibody (OTI4E3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504562S
This item is not returnable.
View return policy
Description
SPINT1 Monoclonal antibody specifically detects SPINT1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| SPINT1 | |
| Monoclonal | |
| 0.82 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96DM3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 29919 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI4E3 | |
| Unconjugated | |
| Rmc1 | |
| HAI; HAI1; HAI-1; hepatocyte growth factor activator inhibitor 1; hepatocyte growth factor activator inhibitor type 1; hepatocyte growth factor activator inhibitor-1; kunitz-type protease inhibitor 1; MANSC2; serine peptidase inhibitor, Kunitz type 1; serine protease inhibitor, Kunitz type 1; Spint1; UNQ223/PRO256 | |
| Full length human recombinant protein of human C18orf8 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction