Learn More
Spiramycin, 90%
Product Code. 15427459
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15427459 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15427459 Supplier Thermo Scientific Alfa Aesar Supplier No. J66194.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
CAS: 8025-81-8 | C43H74N2O14 | 843.065 g/mol
Targets Gram-negative, Gram-positive and some toxoplasma
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 8025-81-8
Molecular Formula C43H74N2O14
Molecular Weight (g/mol) 843.065
MDL Number MFCD01314545
InChI Key ACTOXUHEUCPTEW-ZOTSFZJCSA-N
Synonym spiramycin
PubChem CID 131674974
IUPAC Name 2-[(4R,5S,6S,7R,9R,10R,16R)-6-[(2S,3R,4R,5S,6R)-5-[(2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-10-[(2R,5S,6R)-5-(dimethylamino)-6-methyloxan-2-yl]oxy-4-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-1-oxa
SMILES CC1CC=CC=CC(C(CC(C(C(C(CC(=O)O1)O)OC)OC2C(C(C(C(O2)C)OC3CC(C(C(O3)C)O)(C)O)N(C)C)O)CC=O)C)OC4CCC(C(O4)C)N(C)C
Specifications
Boiling Point 914°C
Flash Point >110°C (230°F)
Packaging Glass bottle
Quantity 1 g
Solubility Information Soluble in methanol. Slightly soluble in water
Percent Purity 90%
Physical Form Powder
Chemical Name or Material Spiramycin