missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Splicing Factor 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00
Specifications
| Antigen | Splicing Factor 1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230031
|
Novus Biologicals
NBP3-38261-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30230743
|
Novus Biologicals
NBP3-38261-100ul |
100 μL |
N/A
|
N/A | |||||
Description
Splicing Factor 1 Polyclonal antibody specifically detects Splicing Factor 1 in Human,Mouse samples. It is validated for ELISA,Western BlotSpecifications
| Splicing Factor 1 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Mammalian branch point-binding protein, mBBP, splicing factor 1, Transcription factor ZFM1, ZFM1BBP, Zinc finger gene in MEN1 locus, Zinc finger protein 162D11S636, ZNF162MBBP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human Splicing Factor 1 (NP_973727.1).,, Sequence:, MATGANATPLDFPSKKRKRSRWNQDTMEQKTVIPGMPTVIPPGLTREQERAYIVQLQIEDLTRKLRTGDLGIPPNPEDRSPSPEPIYNSEGKRLNTREFRTRKKLEEERHNLITEMVALNPDFKPPADYKPPATRVSDKVMIPQDEYPEINFVGLLIGPRGNTLKNIEKECNAKIMIRGKGSVKEGKVGRKDGQMLPGEDEPLHALVTANTMENVKKAVEQIRNILKQGIETPEDQNDLRKMQLRELARLNGTLREDDNRILRPWQSSETRSITNTTVCT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 7536 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title