Learn More
Carbolite™ Split Tube Furnace
Click to view available options
Length (Metric):
350mm
700mm
Wattage:
1000 w
3000 w
Unit Size:
Each
Specifications
Specifications
| Analysis Time | 45 min. |
| Type | Split Tube Furnace |
| Temperature Stability | 1200°C |
| Dimensions (L x W x H) Exterior | 350 W x 700 H mm |
| Length (Metric) | 700mm |
| Voltage | 220 V |
| Wattage | 3000 w |