Learn More
Trajan™ CTC PAL Fixed Needle Syringe
Click to view available options
Needle Gauge:
22 G
23 G
Diameter (Metric) Outer Needle:
0.63 mm
0.72 mm
Needle Point Style:
Cone
Side Hole
Unit Size:
Each
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
Specifications
Specifications
| Description | Gas tight |
| Needle Gauge | 22 G |
| Diameter (Metric) Outer | 0.72 mm |
| Diameter (Metric) Outer Needle | 0.72 mm |
| Product Type | Fixed Needle Syringe |
| Model | 10F-CTC-GT-5/0.72H |
| Quantity | 1 Ea. |
| Volume (Metric) | 10 μL |
| Needle Point Style | Side Hole |