Learn More
Thermo Scientific™ SPME Arrow Liner
Brand:  Thermo Scientific™ 453A0411
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 15840705
  • € 336.27 / Pack of 3
Estimated Shipment: 14-06-2024
to see stock.
Specifications
Triplus RSH Autosampler
SPME Arrow Liner ID 1.3 mm
SPME Arrow