missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPN Monoclonal Antibody (OTI2D4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800786S
This item is not returnable.
View return policy
Description
SPN Monoclonal antibody specifically detects SPN in Human samples. It is validated for Western BlotSpecifications
| SPN | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P16150 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6693 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2D4 | |
| Unconjugated | |
| SPN | |
| 3E8 antigen; A630014B01Rik; B-cell differentiation antigen LP-3; CD43; CD43 cytoplasmic tail; CD43-ct; Galactoglycoprotein; GALGP; gpL115; Leukocyte sialoglycoprotein; leukosialin; leukosianin; LOC101120462; LOW QUALITY PROTEIN: leukosialin; Lsn; Lsn1; Ly48; Ly-48; Lymphocyte antigen 48; sialophorin; sialophorin (gpL115, leukosialin, CD43); sialophorin gpL115; SPN; W3/13 antigen | |
| Full length human recombinant protein of human SPN produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction