Learn More
SPOP Antibody (S01-5B7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19781-50ul
 View more versions of this product
Product Code. 18647663
  • € 421.00 / 50µL
Description
SPOP Monoclonal antibody specifically detects SPOP in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SPOP
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
8405
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot
S01-5B7
Unconjugated
HIB homolog 1, Roadkill homolog 1, speckle-type POZ protein, TEF2
A synthetic peptide of human SPOP (Uniprot # O43791)