missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SPOP Recombinant Rabbit Monoclonal Antibody (23GB4650), Invitrogen™

Product Code. p-200087830
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30089437 100 μL 100µL
30088119 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30089437 Supplier Invitrogen™ Supplier No. MA552722

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Component of a cullin-RING-based BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex that mediates the ubiquitination of target proteins, leading most often to their proteasomal degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPOP
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4650
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SPOP
Gene Accession No. O43791
Gene Alias AI315626; BTBD32; HIB homolog 1; Pcif1; PDX-1 C-terminal-interacting factor 1; roadkill homolog 1; similar to speckle-type POZ protein; speckle type BTB/POZ protein; speckle type POZ protein; speckle-type POZ protein; Spop; SPOP1; TEF2; zgc:64140
Gene Symbols SPOP
Host Species Rabbit
Immunogen A synthesized peptide derived from human SPOP (300-335AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8405
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.