missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPOP Recombinant Rabbit Monoclonal Antibody (2N8L8)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA537921
This item is not returnable.
View return policy
Description
Positive Samples: PC-3 Immunogen sequence: VFKEMMCFIY TGKAPNLDKM ADDLLAAADK YALERLKVMC EDALCSNLSV ENAAEILILA DLHSADQLKT QAVDFINYHA SDVLETSGWK SMVVSHPHLV A.
Component of a cullin-RING-based BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex that mediates the ubiquitination of target proteins, leading most often to their proteasomal degradation.
Specifications
| SPOP | |
| Recombinant Monoclonal | |
| 1.08 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43791 | |
| SPOP | |
| A synthetic peptide corresponding to a sequence within amino acids 250-350 of human SPOP (O43791). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 2N8L8 | |
| Unconjugated | |
| SPOP | |
| AI315626; BTBD32; HIB homolog 1; Pcif1; PDX-1 C-terminal-interacting factor 1; roadkill homolog 1; similar to speckle-type POZ protein; speckle type BTB/POZ protein; speckle type POZ protein; speckle-type POZ protein; Spop; SPOP1; TEF2; zgc:64140 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 8405 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction