Learn More
Haldenwanger™ Spouted Mortar
Click to view available options
Diameter (Metric) Outer:
50mm
70mm
80mm
115mm
180mm
240mm
Unit Size:
Each
Pack of 5
Specifications
Specifications
| Capacity (Metric) | 75 mL |
| Diameter (Metric) Outer | 70mm |
| Product Type | Mortar |
| Includes | With spout |
| Type | Mortar |