Learn More
SPP1 Monoclonal Antibody (OTI6C12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA806238
Encompass_Preferred
 View more versions of this product
Product Code. 17596343
  • € 540.87 / 100µL
Description
Osteopontin is an arginine-glycine-aspartic acid (RGD)-containing glycoprotein that interacts with integrins and CD44 as major receptors. It is a multifunctional protein involved in bone mineralization, cell adhesion, cell migration, chronic inflammatory disease and transformation. Proteolytic cleavage by thrombin and matrix metalloproteinases close to the integrin-binding Arg-Gly-Asp sequence modulates the function of OPN and its integrin binding properties. Thrombin-cleaved fragments of Osteopontin are overexpressed in maligt glial tumors and provide a molecular niche with survival advantage and provide a novel substrate for plasmin and cathepsin D.
Specifications
Specifications
SPP1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P10451
SPP1
Human recombit protein fragment corresponding to amino acids 17-314 of human SPP1 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI6C12
Unconjugated
SPP1
BNSP, BSPI, ETA-1, OPN
Mouse
Affinity Chromatography
RUO
6696
-20° C, Avoid Freeze/Thaw Cycles