missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPPL2A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598784
This item is not returnable.
View return policy
Description
SPPL2A Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot
By searching sequence databases for homologs of Dictyostelium discoideum Impas, which shares homology with presenilin, followed by PCR of lymphocyte and hippocampus cDNA libraries, Grigorenko et al. (2002) cloned SPPL2a. This protein, which may act as an intramembrane protease, contains an N-terminal protease-associated (PA) domain, several transmembrane regions, a hydrophilic loop, conservative sequences around the first and second aspartate residues, and an invariant PAL motif near the C terminus.
Specifications
| SPPL2A | |
| Polyclonal | |
| Unconjugated | |
| SPPL2A | |
| 2010106G01Rik; C130089K23Rik; IMP3; IMP-3; intramembrane cleaving protease; intramembrane protease 3; MNCb-3763; Presenilin-like protein 2; protein SPPL2a; PSEC0147; PSL2; RGD1563001; signal peptide peptidase like 2A; signal peptide peptidase-like 2A; SPPL2A; SPPL2a protein; SPP-like 2A; SPP-like 2A protein | |
| Rabbit | |
| Protein G | |
| RUO | |
| 84888 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8TCT8 | |
| SPPL2A | |
| Recombinant Human Signal peptide peptidase-like 2A protein (26-172AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction