Learn More
MedChemExpress SPQ
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Specifications
Specifications
| Quantity | 50 mg |
| Storage Requirements | 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) |
| Solubility | H2O : 25 mg/mL (88.86 mM; Need ultrasonic) ∣ DMSO : < 1 mg/mL (insoluble or slightly soluble) |
| Shipping Condition | Room temperature |
| CAS | 83907-40-8 |
| Molecular Weight (g/mol) | 281.33 g/mol |
| Target | HBV |